Learn More
ACROBiosystems Biotinylated Human TSLP Protein, His, Avitag™
Biotinylated Protein
441.35 CHF - 1941.94 CHF
Spezifikation
Zugriffsnummer | NP_149024.1 |
---|---|
Molekulargewicht | 18.6 kDa |
Quelle | Biotinylated Human TSLP, His,Avitag (TSP-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1). |
Immunogen | Tyr 29 - Gln 159 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15817877
|
ACROBiosystems
TSP-H82EB-200UG |
200 μg |
1941.94 CHF
200 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827877
|
ACROBiosystems
TSP-H82EB-25UG |
25 μg |
441.35 CHF
25 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Thymic stromal lymphopoietin (TSLP) is an epithelial cell-derived cytokine involved in the pathology of inflammatory skin diseases, and is widely expressed by epithelial cells. Human TSLP cDNA encodes a 159 amino acid (aa) residue precursor protein with a 28 aa signal sequence (4, 5). Human TSLP has been shown to developing nondeletional central tolerance, amplifying epithelium-induced class switching, inducing atopic diseases and maintaining intestinal noninflammatory environment. Among diverse cells responding to Human TSLP, CD11c+ dendritic cells are the most obviously characterized target cells.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.6 kDa. As a result of glycosylation, the protein migrates as 11 kDa and 18-21 kDa under reducing (R) condition, and 22-28 kDa under nSpezifikation
NP_149024.1 | |
Biotinylated Human TSLP, His,Avitag (TSP-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1). | |
-20°C | |
TSLP | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
18.6 kDa | |
Tyr 29 - Gln 159 | |
TSLP | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |