missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human SLAMF7 / CRACC / CD319 Protein, Avitag™,His Tag

Biotinylated Protein

437.70 CHF - 1925.89 CHF

Spezifikation

Zugriffsnummer AAH27867
Molekulargewicht 25.6 kDa
Quelle Biotinylated Human SLAMF7, Avitag,His Tag (SL7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # Q9NQ25-1).
Immunogen Ser 23 - Met 226
Lagerungsbedingungen -20°C
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15827717
Unterlagen ansehen Aktionsdetails
ACROBiosystems
SL7-H82E0-25UG
25 μg
437.70 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15817717
Unterlagen ansehen Aktionsdetails
ACROBiosystems
SL7-H82E0-200UG
200 μg
1925.89 CHF
200 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

SLAM family member 7 (SLAMF7) is also known as CD2-like receptor-activating cytotoxic cells (CRACC), Membrane protein FOAP-12, CD antigen CD319, Novel Ly9, Protein 19A, which is a single-pass type I membrane protein and a member of the CD2 family of cell surface receptors. SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Isoform 1 of SLAMF7 mediates NK cell activation through a SH2D1A-independent extracellular signal-regulated ERK-mediated pathway. May play a role in lymphocyte adhesion. Isoform 3 of SLAMF7 does not mediate any NK cell activation.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

AAH27867
Biotinylated Human SLAMF7, Avitag,His Tag (SL7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # Q9NQ25-1).
-20°C
SLAMF7
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
25.6 kDa
Ser 23 - Met 226
SLAMF7,CD319,CS1,CRACC,19 A,FOAP-12
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by reduced SDS-PAGE
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications