Learn More
ACROBiosystems Biotinylated Human SCF / KITLG Protein, Avitag™,His Tag
Biotinylated Protein
Marke: ACROBiosystems SCF-H82E1-25UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Kit ligand (KITLG) is also known as stem cell factor (SCF), mast cell growth factor (MGF), steel factor (SF), which belongs to the SCF family, and is a widely expressed 28 - 40 kDa type I transmembrane glycoprotein. KITLG is the ligand for the receptor-type protein-tyrosine kinase KIT. SCF / MGF plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. KITLG / SCF binding can activate several signaling pathways. KITLG / SF Promotes phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase, and subsequent activation of the kinase AKT1. KITLG / SCF and KIT also transmit signals via GRB2 and activation of RAS, RAF1 and the MAP kinases MAPK1/ERK2 and/or MAPK3/ERK1. KITLG / SCF and KIT promote activation of STAT family members STAT1, STAT3 and STAT5. KITLG / SCF and KIT promote activation of PLCG1, leading to the production of the cellular signaling molecules diacylglycerol and inositol 1, 4, 5 - trisphosphate. KITLG / SCF acts synergistically with other cytokines, probably interleukins.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 21.7 kDa. The protein migrates as 26-38 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
AAH69797 | |
25 μg | |
Glu 26 - Ala 190 | |
KITLG,FPH2,KL-1,Kitl,MGF,SCF,SF,SHEP7,KL | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
21.7 kDa | |
Biotinylated Human SCF, Avitag,His Tag (SCF-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Ala 190 (Accession # AAH69797). | |
-20°C | |
SCF | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |