Learn More
ACROBiosystems Biotinylated Human PD-L2 / B7-DC Protein, Fc, Avitag™
Biotinylated Protein
441.35 CHF - 1941.94 CHF
Spezifikation
Zugriffsnummer | AAI13679 |
---|---|
Molekulargewicht | 51.0 kDa |
Quelle | Biotinylated Human PD-L2, Fc,Avitag (PD2-H82F6) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # Q9BQ51-1). |
Immunogen | Leu 20 - Pro 219 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15817517
|
ACROBiosystems
PD2-H82F6-200UG |
200 μg |
1941.94 CHF
200 Mikrogramm |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827517
|
ACROBiosystems
PD2-H82F6-25UG |
25 μg |
441.35 CHF
25 Mikrogramm |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Programmed cell death 1 ligand 2 (PD-L2 or PDCD1 ligand 2) is also known as Butyrophilin B7-DC, CD antigen CD273, which belongs to the immunoglobulin superfamily or BTN/MOG family. The expression of PD-L2 is up-regulated by IFNG/IFN-gamma stimulation in monocytes and induced on dendritic cells grown from peripheral blood mononuclear cells with CSF2 and IL-4. PD-L2 Involved in the costimulatory signal, essential for T-cell proliferation and IFNG production in a PDCD1-independent manner. PD-L2 interaction with PDCD1 inhibits T-cell proliferation by blocking cell cycle progression and cytokine production.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.0 kDa. The protein migrates as 66-91 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAI13679 | |
Biotinylated Human PD-L2, Fc,Avitag (PD2-H82F6) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # Q9BQ51-1). | |
-20°C | |
PD-L2 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
51.0 kDa | |
Leu 20 - Pro 219 | |
PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |