missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human PD-L1 / B7-H1 Protein, Avitag™,His Tag (recommended for biopanning, MALS verigied)

Biotinylated Protein

512.74 CHF - 2213.53 CHF

Spezifikation

Zugriffsnummer NP_054862.1
Molekulargewicht 27.8 kDa
Quelle Biotinylated Human PD-L1, Avitag™,His Tag (recommended for biopanning) (PD1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).
Immunogen Phe 19 - Arg 238
Lagerungsbedingungen -20°C
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15827477
Unterlagen ansehen Aktionsdetails
ACROBiosystems
PD1-H82E5-25UG
25 μg
512.74 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15817477
Unterlagen ansehen Aktionsdetails
ACROBiosystems
PD1-H82E5-200UG
200 μg
2213.53 CHF
200 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 27.8 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

NP_054862.1
Biotinylated Human PD-L1, Avitag™,His Tag (recommended for biopanning) (PD1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).
-20°C
PD-L1
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
27.8 kDa
Phe 19 - Arg 238
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications