Learn More
ACROBiosystems Biotinylated Human LILRA5 / CD85f / ILT11 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems LI5-H82E1-200UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
LIRs are immunoglobulin-like receptors that have activating and inhibitory functions in leukocytes. Leukocyte immunoglobulin-like receptor A5 (LILRA5) belongs to a family of receptors known to regulate leukocyte activation. LILRA5, expressed as a membrane-bound receptor and as a secreted molecule. The transmembrane LILRA5 contain a short cytoplasmic domain and a charged arginine residue within the transmembrane region that is likely to mediate its association with another coreceptor. LILRA5 is mostly expressed in myeloid cells, including monocytes and neutrophils.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_871714.1 | |
200 μg | |
Gly 42 - Arg 268 | |
LILRA5,CD85f,ILT11,LIR9,LILRB7 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
28.9 kDa | |
Biotinylated Human LILRA5, His,Avitag (LI5-H82E1) is expressed from human 293 cells (HEK293). It contains AA Gly 42 - Arg 268 (Accession # A6NI73-1). | |
-20°C | |
LILRA5 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |