Learn More
ACROBiosystems Biotinylated Human LILRA2 / CD85h / ILT1 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems LI2-H82E9-25UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
LILRA2 (Leukocyte immunoglobulin like receptor A2), also known as CD85H and LIR7 (Leukocyte immunoglobulin-like receptor 7). It belongs to the subfamily A class of LIR receptors (LILRAs), which are single-pass type I transmembrane proteins with 2-4 extracellular Ig-like domains, a transmembrane domain (TM), and a short cytoplasmic tail. LILRA2 contains 4 Ig-like C2 type domains in the extracellular region. LILRA2 does not bind class I MHC antigens. LILRA2 is expressed predominantly on monocytes and B cells, and at lower levels on dendritic cells and natural killer cells. LILRA2 is an activating receptor that inhibits dendritic cell differentiation and antigen presentation and suppresses innate immune response.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
Q8N149.2 | |
25 μg | |
Gly 24 - Asn 449 | |
ILT1,LIR7,CD85H,LIR-7,LILRA2,ILT1CD85H,CD85h,ILT1,ILT-1,ILT1CD85H,CD85h antigen,CD85 antigen-like family member H | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
50.7 kDa | |
Biotinylated Human LILRA2, His,Avitag (LI2-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 449 (Accession # Q8N149-1). | |
-20°C | |
LILRA2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |