Learn More
ACROBiosystems Biotinylated Human IL-17 RA / CD217 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems ILR-H82E5-25UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Interleukin-7 receptor subunit alpha (IL-7 RA or IL7Ra) is also known as cluster of differentiation 127 (CD127). IL-7 RA is a type I cytokine receptor and is a subunit of the functional Interleukin-7 receptor and thymic stromal lymphopoietin (TSLP) receptors. IL-7 RA has been shown to play a critical role in the development of immune cells called lymphocytes - specifically in a process known as V(D)J recombination[citation needed]. This protein is also found to control the accessibility of a region of the genome that contains the T-cell receptor gamma gene, by STAT5 and histone acetylation.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.2 kDa. The protein migrates as 55-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_055154.3 | |
25 μg | |
Leu 33 - Trp 320 | |
CD217,CDw217,IL-17 RA,IL17R,CANDF5,hIL-17 R | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
37.2 kDa | |
Biotinylated Human IL-17 RA, His,Avitag (ILR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1). | |
-20°C | |
IL-17 RA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |