Learn More
ACROBiosystems Biotinylated Human IL-17 RA / CD217 Protein, His, Avitag™
Biotinylated Protein
Marke:  ACROBiosystems ILR-H82E5-200UG
Additional Details : Gewicht : 0.00001kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15827127
  • 1941.94 CHF / 200 Mikrogramm
Verfügbar ab: 13-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Interleukin-7 receptor subunit alpha (IL-7 RA or IL7Ra) is also known as cluster of differentiation 127 (CD127). IL-7 RA is a type I cytokine receptor and is a subunit of the functional Interleukin-7 receptor and thymic stromal lymphopoietin (TSLP) receptors. IL-7 RA has been shown to play a critical role in the development of immune cells called lymphocytes - specifically in a process known as V(D)J recombination[citation needed]. This protein is also found to control the accessibility of a region of the genome that contains the T-cell receptor gamma gene, by STAT5 and histone acetylation.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.2 kDa. The protein migrates as 55-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_055154.3
200 μg
Leu 33 - Trp 320
CD217,CDw217,IL-17 RA,IL17R,CANDF5,hIL-17 R
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
37.2 kDa
Biotinylated Human IL-17 RA, His,Avitag (ILR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1).
-20°C
IL-17 RA
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins