Learn More
ACROBiosystems Biotinylated Human IgG1 Fc protein, Avitag™
Biotinylated Protein
Marke: ACROBiosystems IG1-H8213-25UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.
This protein carries an Avi tag at the C-terminus. The protein has a calculated MW of 28.4 kDa. The protein migrates as 32-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
BAC85237.1 | |
25 μg | |
Glu 99 - Lys 330 | |
IgG1 | |
Precision Avi | |
Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
28.4 kDa | |
Biotinylated Human IgG1 Fc, Avitag (IG1-H8213) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857-1). | |
-20°C | |
IgG1 Fc | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |