missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human IGF-II Protein, Avitag™,Fc Tag

Biotinylated Protein

437.70 CHF - 1925.89 CHF

Spezifikation

Molekulargewicht 35.3 kDa
Quelle Biotinylated Human IGF-II, Avitag,Fc Tag (IG2-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession # P01344-1).
Immunogen Ala 25 - Glu 91
Lagerungsbedingungen -20°C
Gen-Alias IGF2,C11orf43,FLJ22066,FLJ44734,IGF-II,PP9974
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15836997
Unterlagen ansehen Aktionsdetails
ACROBiosystems
IG2-H82F9-25UG
25 μg
437.70 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15826997
Unterlagen ansehen Aktionsdetails
ACROBiosystems
IG2-H82F9-200UG
200 μg
1925.89 CHF
200 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Insulin-like growth factor 2 (IGF-2) is also known as Somatomedin-A, IGF-II, PP9974, and is one of three protein hormones that share structural similarity to insulin. IGF-2 exerts its effects by binding to the IGF-1 receptor. IGF2 may also bind to the IGF-2 receptor (also called the cation-independent mannose 6-phosphate receptor), which acts as a signalling antagonist; that is, to prevent IGF2 responses. The major role of IGF2 is as a growth promoting hormone during gestation. In the process of Folliculogenesis, IGF2 is created by Theca cells to act in an autocrine manner on the theca cells themselves, and in a paracrine manner on Granulosa cells in the ovary. IGF2 promotes granulosa cell proliferation during the follicular phase of the menstrual cycle, acting alongside Follicle Stimulating Hormone (FSH). After ovulation has occurred, IGF-2 promotes progesterone secretion during the luteal phase of the menstrual cycle together with Luteinizing Hormone (LH). Thus, IGF2 acts as a Co-hormone together with both FSH and LH. IGF-2 may play a key role in memory and could potentially be used to treat Alzheimer's Disease. It is sometimes produced in excess in islet cell tumours, causing hypoglycemia. Doege-Potter syndrome is a paraneoplastic syndrome in which hypoglycemia is associated with the presence of one or more non-islet fibrous tumors in the pleural cavity. has been shown to interact with IGFBP3 and Transferrin.

This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 35.3 kDa. As a result of glycosylation, the protein migrates as 40 kDa under reducing (R) condition, and 66 kDa under non-redu
Spezifikation

Spezifikation

35.3 kDa
Ala 25 - Glu 91
IGF2,C11orf43,FLJ22066,FLJ44734,IGF-II,PP9974
Precision Avi
Avi Tag and Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Biotinylated Human IGF-II, Avitag,Fc Tag (IG2-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession # P01344-1).
-20°C
IGF-II
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
MABSol™ Biotin Labeled Proteins
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications