Learn More
ACROBiosystems Biotinylated Human ErbB3 / Her3 Protein, His Tag, ultra sensitivity (primary amine labeling)
365.69 CHF - 1752.79 CHF
Spezifikation
Zugriffsnummer | NP_001973.2 |
---|---|
Molekulargewicht | 69.5 kDa |
Quelle | MABSol™ Biotinylated Human ErbB3, His Tag, primary amine labeling (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2). It is the biotinylated form of Human ErbB3, His Tag (ER3-H5223). |
Immunogen | Ser 20 - Thr 643 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15826687
|
ACROBiosystems
ER3-H8223-200UG |
200 μg |
1752.79 CHF
200 Mikrogramm |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15836687
|
ACROBiosystems
ER3-H8223-25UG |
25 μg |
365.69 CHF
25 Mikrogramm |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
ErbB3, also known as Her3 (human epidermal growth factor receptor 3), is a member of the epidermal growth factor receptor (EGFR) family of receptor tyrosine kinases. This membrane-bound glycoprotein has a neuregulin binding domain but has not an active kinase domain. It therefore can bind the ligand but cannot mediate the intracellular signal transduction through protein phosphorylation. However, it does form heterodimers with ErbB2 or other EGFR members responsible for tyrosine phosphorylation to give a receptor complex and initiate the related pathway, which lead to cell proliferation or differentiation. Overexpression of this protein has been reported in numerous cancers, including prostate, bladder, and breast tumors. This protein has different isoforms derived from alternative splicing variants, and among which, the secreted isoform lacking the intermembrane region modulates the activity of membrane-bound form.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 69.5 kDa. The protein migrates as 85-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001973.2 | |
MABSol™ Biotinylated Human ErbB3, His Tag, primary amine labeling (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2). It is the biotinylated form of Human ErbB3, His Tag (ER3-H5223). | |
-20°C | |
ErbB3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
69.5 kDa | |
Ser 20 - Thr 643 | |
ERBB3,HER3,LCCS2,MDA-BF-1,MGC88033,c-erbB3,erbB3-S,p180-ErbB3,p45-sErbB3,p85-sErbB3 | |
UltraLys | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |