Learn More
ACROBiosystems Biotinylated Human EpCAM / TROP1 Protein, His Tag, ultra sensitivity (primary amine labeling)
365.69 CHF - 1588.86 CHF
Spezifikation
Zugriffsnummer | AAH14785.1 |
---|---|
Molekulargewicht | 28.2 kDa |
Quelle | MABSol™ Biotinylated Human EpCAM, His Tag, primary amine labeling (EPM-H8223) is expressed from human HEK293 cells. It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1). It is the biotinylated form of Human EpCAM, His Tag (EPM-H5223). |
Immunogen | Gln 24 - Lys 265 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15826667
|
ACROBiosystems
EPM-H8223-25UG |
25 μg |
365.69 CHF
25 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15816667
|
ACROBiosystems
EPM-H8223-200UG |
200 μg |
1588.86 CHF
200 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
EpCAM is also known as CO171A, EGP, EGP40,GA7332, KSA, M4S, MIC18, MK1, TROP1, hEGP2, and is a pan-epithelial differentiation antigen that is expressed on almost all carcinomas as 17-1A(mAb) antigen. Its constitutional function is being elucidated. It is intricately linked with the Cadherin-Catenin pathway and hence the fundamental WNT pathway responsible for intracellular signaling and polarity. The epithelial cell adhesion molecule (Ep-CAM) is known to express in most epithelial malignancies and was reported as a tumor marker or a candidate of molecular targeting therapy. Ep-CAM cross signaling with N-cadherin involves Pi3K, resulting in the abrogation of the cadherin adhesion complexes in epithelial cells was reported. And Epithelial cell adhesion molecule (Ep-CAM) recently received increased attention as a prognostic factor in breast cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 33-36 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH14785.1 | |
MABSol™ Biotinylated Human EpCAM, His Tag, primary amine labeling (EPM-H8223) is expressed from human HEK293 cells. It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1). It is the biotinylated form of Human EpCAM, His Tag (EPM-H5223). | |
-20°C | |
EpCAM | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
28.2 kDa | |
Gln 24 - Lys 265 | |
EPCAM,TACSTD1,TROP1,CD326,DIAR5,EGP2,EGP314,EGP40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1/4,KSA,M4S1,MIC18,MK1 | |
UltraLys | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |