Learn More
ACROBiosystems Biotinylated Human CEACAM-1 / CD66a Protein, Avitag™,His Tag
Biotinylated Protein
Marke: ACROBiosystems CE1-H82E5-25UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) is also known as Biliary glycoprotein 1 (BGP1), CD66a, which belongs to the immunoglobulin superfamily or CEA family. CEACAM1 /CD66a contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CEACAM1 /CD66a was described as an adhesion molecule mediating cell adhesion via both homophilic and heterophilic manners, and was detected on leukocytes, epithelia, and endothelia. Studies have revealed that CEACAM1 / BGP-1 performs actions in multiple cellular processes including tissue differentiation, angiogenesis, apoptosis, metastasis, as well as the modulation of innate and adaptive immune responses.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 45.9 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001703.2 | |
25 μg | |
Gln 35 - Gly 428 | |
CEACAM1,CD66a,BGP,BGP1,BGPI | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |
45.9 kDa | |
Biotinylated Human CEACAM-1, Avitag,His Tag (CE1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # P13688-1). | |
-20°C | |
CEACAM-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |