Learn More
ACROBiosystems Biotinylated Human CEACAM-1 / CD66a Protein, Avitag™,His Tag
Biotinylated Protein
Marke:  ACROBiosystems CE1-H82E5-200UG
Additional Details : Gewicht : 0.00001kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15826437
  • 1941.94 CHF / 200 Mikrogramm
Verfügbar ab: 14-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) is also known as Biliary glycoprotein 1 (BGP1), CD66a, which belongs to the immunoglobulin superfamily or CEA family. CEACAM1 /CD66a contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CEACAM1 /CD66a was described as an adhesion molecule mediating cell adhesion via both homophilic and heterophilic manners, and was detected on leukocytes, epithelia, and endothelia. Studies have revealed that CEACAM1 / BGP-1 performs actions in multiple cellular processes including tissue differentiation, angiogenesis, apoptosis, metastasis, as well as the modulation of innate and adaptive immune responses.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 45.9 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001703.2
200 μg
Gln 35 - Gly 428
CEACAM1,CD66a,BGP,BGP1,BGPI
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by reduced SDS-PAGE
45.9 kDa
Biotinylated Human CEACAM-1, Avitag,His Tag (CE1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # P13688-1).
-20°C
CEACAM-1
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins