Learn More
ACROBiosystems Biotinylated Human CD30 / TNFRSF8 Protein, Avitag™,His Tag
MALS verified
441.35 CHF - 1941.94 CHF
Spezifikation
Zugriffsnummer | NP_001234.2 |
---|---|
Molekulargewicht | 41.1 kDa |
Quelle | Biotinylated Human CD30, Avitag,His Tag (CD0-H82E6) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). |
Immunogen | Phe 19 - Lys 379 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15816137
|
ACROBiosystems
CD0-H82E6-200UG |
200 μg |
1941.94 CHF
200 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826137
|
ACROBiosystems
CD0-H82E6-25UG |
25 μg |
441.35 CHF
25 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Human CD30 is also known as TNFRSF8, is a cell membrane protein of the tumor necrosis factor receptor family and tumor marker. TNFRSF-8 is expressed by activated, but not by resting, T and B cells. Also, CD30 is expressed on classical Hodgkin Lymphoma cells together with CD15. CD30 is the receptor for TNFSF8/CD30L. CD30 can interact with TRAF2 and TRAF5, and mediate the signal transduction that leads to the activation of NF-kappa-B. TNFRSF8 may play a role in the regulation of cellular growth and transformation of activated lymphoblasts. TNFRSF8 is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 41.1 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001234.2 | |
Biotinylated Human CD30, Avitag,His Tag (CD0-H82E6) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). | |
-20°C | |
CD30 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
41.1 kDa | |
Phe 19 - Lys 379 | |
TNFRSF8,CD30,D1S166E,Ki-1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |