Learn More
ACROBiosystems Biotinylated Human B7-H7 / HHLA2 Protein, Fc, Avitag™
Biotinylated Protein
441.35 CHF - 1941.94 CHF
Spezifikation
Zugriffsnummer | NP_009003.1 |
---|---|
Molekulargewicht | 65.9 kDa |
Quelle | Biotinylated Human B7-H7, Fc,Avitag (B77-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1). |
Immunogen | Ile 23 - Asn 344 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15835987
|
ACROBiosystems
B77-H82F5-25UG |
25 μg |
441.35 CHF
25 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15825987
|
ACROBiosystems
B77-H82F5-200UG |
200 μg |
1941.94 CHF
200 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
B7-H7 (HHLA2) is a newly identified B7 family member that regulates human T-cell functions. B7-H7 was previously known as human endogenous retrovirus-H long terminal repeat associating 2 (HHLA2) with unidentified function. Recently, B7-H7 has been identified as a specific ligand for human CD28H. The B7-H7-CD28H pathway strongly promoted CD4+ T-cell proliferation and cytokine production via an AKT-dependent signaling cascade in the presence of TCR signaling, suggesting B7-H7 comprises a new co-stimulatory pathway. The first IgV domain of B7-H7, which presumably binds to a putative receptor, shows the highest homology to other B7 family members.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 65.9 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_009003.1 | |
Biotinylated Human B7-H7, Fc,Avitag (B77-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1). | |
-20°C | |
B7-H7 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
65.9 kDa | |
Ile 23 - Asn 344 | |
B7-H7,HHLA2,B7 Homolog 7 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |