Learn More
Merck Bioprocess Vantage Chromatography Column
Vantage-L laboratory columns provide advanced design for the increasing demands of biochromatography
Marke: Merck 96440250
Additional Details : Gewicht : 1.00000kg
Beschreibung
Vantage-L laboratory columns provide advanced design for the increasing demands of biochromatography. Five diameters and three lengths provide a range of column volumes and bed heights to select from, with the twin adjusters providing 250mm adjustment in all columns. A 7bar pressure rating and resistance to commonly used solvents makes them ideally suited to smaller particle media applications.Minimal components and simple but positive adjuster positioning, ensure ease of use. Zero deadspace seal design, coupled with the proven Millipore flow distributor, provides optimal chromatographic efficiency which is scalable to pilot and production columns.Spezifikation
44 mm | |
250 mm | |
15.2cm2 |
Chromatography Column | |
40°C |