missing translation for 'onlineSavingsMsg'
Learn More

Bevantolol Hydrochlorid, TRC

Product Code. 30006982
Change view
Click to view available options
Menge:
10 mg
Unit Size:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30006982 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006982 Supplier Toronto Research Chemicals Supplier No. TRCB33450010MG
 Eingeschränkter Artikel
Verfügbar ab: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42864-78-8
Summenformel C20H28ClNO4
Molekulargewicht (g/mol) 381.89
Synonym 1-[[2-(3,4-Dimethoxyphenyl)ethyl]amino]-3-(3-methylphenoxy)-2-propanol Hydrochloride, Dimethoxyphenethyl)amino]-3-(m-tolyloxy)-2-propanol Hydrochloride, CI 775, Cl 775, NC 1400, Ranestol, Sentiloc, Vantol;
SMILES Cl.COc1ccc(CCNCC(O)COc2cccc(C)c2)cc1OC

Specifications

Chemischer Name oder Material Bevantolol Hydrochloride
Menge 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.