missing translation for 'onlineSavingsMsg'
Learn More

Beta-Morpholinopropionitril, TRC

Product Code. 30478646
Change view
Click to view available options
Menge:
1 g
Unit Size:
1 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30478646 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478646 Supplier Toronto Research Chemicals Supplier No. TRCM7238501G
 Eingeschränkter Artikel
Verfügbar ab: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material beta-Morpholinopropionitrile
CAS 4542-47-6
Summenformel C7H12N2O
Menge 1 g
Synonym 3-Morpholinopropanenitrile, 3-Morpholinopropionitrile, N-(2-Cyanoethyl)morpholine, NSC 20547, NSC 41718, 4-Morpholinepropanenitrile
InChI-Formel InChI=1S/C7H12N2O/c8-2-1-3-9-4-6-10-7-5-9/h1,3-7H2
SMILES N#CCCN1CCOCC1
IUPAC-Name 3-Morpholin-4-ylpropanenitril
Molekulargewicht (g/mol) 140.18
Formelmasse 140.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.