missing translation for 'onlineSavingsMsg'
Learn More

Beta-Carotin, TRC

Product Code. 17918334
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
100 mg
missing translation for 'unitSize':
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge missing translation for 'unitSize'
17918334 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 17918334 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC184250100MG
 Eingeschränkter Artikel
Verfügbar ab: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7235-40-7
Summenformel C40 H56
Molekulargewicht (g/mol) 536.87
Synonym β-Carotene, all-trans- (8CI), (all-E)-1,1'-(3,7,12,16-Tetramethyl-1,3,5,7,9,11,13,15,17-octadecanonaene-1,18-diyl)bis[2,6,6-trimethylcyclohexene], BetaVit, Betacarotene, C.I. 40800, C.I. Food Orange 5, Carofertin, Carotaben, Caroten Base 35468, Carotene Base 80, Carotene Base 80S, Carotene Base 80SV, CoroCare, Cyclohexene, 1,1'-(3,7,12,16-tetramethyl-1,3,5,7,9,11,13,15,17-octadecanonaene-1,18-diyl)bis[2,6,6-trimethyl-, (all-E)-, Food Orange 5, KPMK, Lucaratin, Lucarotin, Lucarotin 10CWD/O, Lucarotin 20MCT, Lucarotin 30SUN, Lurotin, NSC 62794, Provatene, Provatenol, Rovimix β-carotene, Serlabo, Solatene, all-E-β-Carotene, all-trans-β-Carotene, β-Carotene
IUPAC-Name 1,3,3-Trimethyl-2-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyl-18-(2,6,6-Trimethylcyclohexen-1-yl)Octadeca-1,3,5,7,9,11,13,15,17-Nonaenyl]Cyclohexen
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C)\C=C\C=C(/C)\C=C\C2=C(C)CCCC2(C)C

Specifications

Chemischer Name oder Material beta-Carotene
Menge 100 mg
InChI-Formel InChI=1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-22,25-28H,15-16,23-24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+
Formelmasse 536.4382
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.