missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta-3 Defensin Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Marke: Invitrogen™ 900-K210
Additional Details : Gewicht : 0.03500kg
Beschreibung
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Spezifikation
P81534 | |
Lot-specific | |
ELISA | |
414325 | |
This Human BD-3 ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate. | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hr 45 min |
Lot-specific | |
Biotin | |
Colorimetric Microplate Reader | |
DEFB103A | |
HRP | |
RUO | |
Human | |
24 hr 30 min |