missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEND5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20931-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
BEND5 | |
Polyclonal | |
Unconjugated | |
BEND5 | |
BEN domain containing 5, BEND5, C1orf165 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67621, 79656 | |
Antibody | |
IgG |
Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7L4P6, Q8C6D4 | |
Bend5 | |
BEND5 Fusion Protein Ag14930 | |
150 μL | |
Primary | |
Human, Mouse | |
Liquid |