Learn More
behr Labor-Technik™ behrotest™ Wasserproben-Homogenisator
Click to view available options
Volumen (metrisch):
25 l
Packungsgröße:
1 Stück
Beschreibung
- Can be carried by two handles
- Is protected in the in top container against damage
- Easy filling with flattened drain insert.
- Scales for exact measurement of sample quantities
- Directly ready for use by inserting the transport rack on the magnetic stirrer
- The speed of the magnetic stirrer can be adjusted by means of a regulator.
- Directly ready for use by inserting the transport rack on the magnetic stirrer
- Scales for exact measurement of sample quantities
- Easy filling with flattened drain insert.
- Is protected in the in top container against damage
Spezifikation
Spezifikation
| Zertifizierung/Konformität | DIN 38402 A 30 |
| Beschreibung | DIN 38402 A 30 |
| Volumen (metrisch) | 25 l |
| Typ | Homogenisierungsgerät |