Learn More
BDKRB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA562254
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FSPWKISMFL SVREDSVPTT ASFSADMLNV TLQGPTLNGT FAQSKCPQVE WLGWLNTIQP Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 47%.
Bradykinin B2 receptor (BDKRB2) expression has been reported in blood, brain, eye, kidney, liver, lung, prostate, skin, small intestine, synovium, uterus, and vessel. ESTs have been isolated from colon, connective tissue, heart, heart/melanocyte/uterus, and kidney libraries. It is associated with G-proteins that activate a phosphatidylinositol-calcium second messenger system.Spezifikation
BDKRB2 | |
Polyclonal | |
Unconjugated | |
BDKRB2 | |
B(2); B2; B2 bradykinin receptor; B2BKR; B2BRA; B2R; BDKRB2; BK2; BK-2; BK-2 receptor; BK2R; BK-2R; BK-2Rs; BKR2; bradykinin receptor B2; bradykinin receptor, beta 2; BRB2; DKFZp686O088; kinin B2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P30411 | |
BDKRB2 | |
Recombinant protein corresponding to Human B2R | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only