Learn More
BD Vacutainer™ Winged Safety Push Button Blood Collection Sets without Luer Adapter
Artikelnummer. p-5237562
Click to view available options
Kanülen-Gauge:
23 G
Packungsgröße:
50 Stück
Artikelnummer. 12547876
Marke: BD 367324
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Blood collection set offers in-vein safety activation provides split-second protection against accidental needlestick injuries
  • Ease of use reduces training requirements and improves activation compliance
  • In-vein activation reduces the risk exposure to a contaminated needle
  • Easy activation without additional patient discomfort
  • Ideal for use in high-risk environments No luer adapter
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Per Knopfdruck aktiviertes Blutentnahme-Set
Kanülen-Gauge 23 G
Länge (angloamerikanisch) Kanüle 3/4 in.
Länge (angloamerikanisch) Röhrchen 12 in.