missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Serum Tellurite Agar
Beschreibung
- Gebrauchsfertig
- Platten im Format 100 x 10 mm und 100 x 15 mm
Spezifikation
Spezifikation
| Zertifizierung/Konformität | ISO/DIS, 11014 |
| Farbe | Hellbeige |
| Form | Flüssig |
| Herstellungsort | USA |
| Medientyp | Serum-Tellurit-Agar |
| Zusammensetzung | Proteose Pepton Nr. 3 20 g/l, Natriumchlorid 5 g/l, Agar 20 g/l, Lammserum 50 ml, Kaliumtelluritlösung 1 % 10 ml |
| Zur Verwendung mit (Anwendung) | Corynebacterium diphtheriae |
| Verpackungsart | Platte |
| Menge | 20 Stück/Packung |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?