missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL7B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11740-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1. Involved in cell cycle progression, maintece of the nuclear structure and stem cell differentiation. May play a role in lung tumor development or progression.Spezifikation
BCL7B | |
Polyclonal | |
Unconjugated | |
BCL7B | |
B cell CLL/lymphoma 7B, BCL7B, Hom s 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12054, 368001, 9275 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q921K9, Q9BQE9 | |
Bcl7b | |
BCL7B Fusion Protein Ag2333 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |