missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bacillus thuringiensis subsp. Kurstaki Cry1Ac Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA5117608
Additional Details : Gewicht : 0.01500kg
Beschreibung
This antibody has been tested in direct-ELISA
Promotes colloidosmotic lysis by binding to the midgut epithelial cells of many lepidopteran larvae.Spezifikation
Bacillus thuringiensis subsp. Kurstaki Cry1Ac | |
Polyclonal | |
Unconjugated | |
Crystaline entomocidal protoxin; Pesticidal crystal protein Cry1Ac | |
Recombinant Bacillus thuringiensis subsp. kurstaki Pesticidal crystal protein Cry1Ac protein (972-1178aa) | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
3 mg/mL | |
10mM PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |