missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ B7-H6 Monoclonal Antibody (JAM1EW), PE, eBioscience™, Invitrogen™

Product Code. 16370624
Change view
Click to view available options
Menge:
100 Tests
25 Tests
Unit Size:
100 Stück
25 Stück
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16370624 25 Tests 25 Stück
16380624 100 Tests 100 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16370624 Supplier Invitrogen™ Supplier No. 12652641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: This JAM1EW monoclonal antibody recognizes human B7-H6 (NCR3LG1) protein on the surface of cells. It recognizes protein on live but not on paraformaldehyde-fixed cells. The JAM1EW clone is specific to B7-H6. When tested in flow cytometry against cells transfected with either B7-H4, B7-H5, B7-H6, or B7-H7, clone JAM1EW only stains positively on B7-H6 transfected cells. Applications Reported: This JAM1EW antibody has been reported for use in flow cytometric analysis. Applications Tested: This JAM1EW antibody has been pre-diluted and tested by flow cytometric analysis of HeLa and HL-60 cell line. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

B7-H6 (NCR3LG1) is a transmembrane endogenous ligand expressed on the surfaces of tumor cells. B7-H6 binds with NKp30 present on NK cells. Binding of B7-H6 to NKp30 causes the ligation of NKp30 subsequently inducing NK cell activation and target cell cytolysis. B7-H6 is not constitutively expressed on normal tissue. Whether it can be expressed endogenously under certain conditions is under investigation. B7-H6 has been detected on circulating pro-inflammatory CD14(+)CD16(+) monocytes in some patients with sepsis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-H6
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon JAM1EW
Konzentration 5 μL/Test
Konjugat PE
Zusammensetzung PBS with BSA and 0.09% sodium azide; pH 7.2
Gen NCR3LG1
Gen-Zugriffsnummer Q68D85
Gen-Alias B7 homolog 6; B7H6; B7-H6; DKFZp686I21167; DKFZp686O24166; natural cytotoxicity triggering receptor 3 ligand 1; natural killer cell cytotoxicity receptor 3 ligand 1; NCR3LG1; putative Ig-like domain-containing protein DKFZp686O24166/DKFZp686I21167
Gensymbole NCR3LG1
Wirtsspezies Mouse
Immunogen B7-H6 recombinant protein.
Reinigungsverfahren Affinity chromatography
Menge 25 Tests
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 374383
Zielspezies Human
Inhalt und Lagerung 4°C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.