Learn More
B7-H4 Monoclonal Antibody (188), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
160.00 CHF - 467.00 CHF
Spezifikation
Antigen | B7-H4 |
---|---|
Klon | 188 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15269209
|
Affymetrix eBioscience
12-5972-82 |
100 μg |
467.00 CHF
100 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15259209
|
Affymetrix eBioscience
12-5972-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The 188 monoclonal antibody was generated against and reacts with mouse B7-H4 also known as B7S1, B7X. Cross reactivity of this antibody to other proteins has not been determined. B7-H4 is a newly discovered member of the B7 family reported to inhibit T cell activation, cell cycle progression and IL-2 production. The ligand for B7-H4 has not been identified yet. Simultaneous double staining of cells with two anti-mouse B7-H4 antibodies, Clone 9 and 188, suggests that epitopes recognized by these mAbs are different and/or there is no steric hindrance when antibodies are used together. 188 stains mouse B7-H4 transfected cells and not spleen cells. Exact expression pattern of B7-H4 has not been fully characterized. Applications Reported: The 188 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 188 antibody has been tested by flow cytometric analysis of transfected cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
B7-H4 (B7x, VTCN1) a costimulatory protein which is reported to function as a negative regulator of T-cell mediated immunity. Although B7-H4 binds an unknown receptor, it is thought to deliver an inhibitory signal to T-cells preventing their proliferation, cell cycle progression and interleukin-2 production. B7-H4 deficient mice are only minimally affected; suggesting B7-H4 is important in the fine tuning of the T-cell mediated immune response. B7-H4 is expressed on activated T-cells, B-cells, monocytes and dendritic cells. Aberrant expression has been associated with cancers of the lung, breast and ovary in humans. B7-H4 is expressed primarily on the membrane of lymphoid cells, and as an immunoinhibitory protein that interacts with receptors on the surface of T lymphocytes, it is involved in mediating cellular and humoral immune responses. Overexpression of B7-H4 is associated with certain malignancies, including ovarian and breast cancer, which may be a mechanism by which tumor cells suppress T cell immunity and facilitate tumor progression. Further, VTCN1 may be a useful biomarker for the early detection of ovarian cancer and Renal Cell Carcinoma (RCC).Spezifikation
B7-H4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q7TSP5 | |
Vtcn1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Vtcn1 |
188 | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
242122 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.