Learn More
Invitrogen™ B3GNT7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5139750
Beschreibung
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KWLDIYCPHV PFIFKGDDDV FVNPTNLLEF LADRQPQENL FVGDVLQHAR PIRRKDNKYY IPGALYGKAS YPPYAGGGGF LMAGSLARRL HHACDTLELY PIDDVFLGMC LEVLGVQPTA HEGFKTFGIS RNRNSRMNKE PCF Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.
B3GNT7 is a glycosyltransferase which may play a role in elongation of the keratan sulfate carbohydrate backbone.
Spezifikation
| B3GNT7 | |
| Polyclonal | |
| Unconjugated | |
| B3GNT7 | |
| B3GNT7; beta 1,3-N-acetylglucosaminyltransferase 7; beta-1,3-Gn-T7; beta-1,3-N-acetylglucosaminyltransferase 7; beta3GnT7; beta-3GnT7; beta3Gn-T7; BGnT-7; C330001H22Rik; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFL0 | |
| B3GNT7 | |
| Recombinant protein corresponding to Human B3GNT7. Recombinant protein control fragment (Product #RP-108725). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.