Learn More
BOLA B.safe Direct Filling System
Beschreibung
- Highly specialized system for the disposal of especially viscous liquids or liquid waste with solid contents
- b.safe Funnel 130 made of static dissipative PE-EX with GL 45 cap, hinged lid and sieve
- Safety base cabinet T90 in colour RAL7035 (light gray), outer dimensions: W 593 x D 574 x 600 mm, all passages in fire protection compliant design
- Telescopic disposal tube made of stainless steel for working height 90 cm with lock nut, canister lid, capacitive fill
- Level sensor, vent hose and collecting vessel
- b.safe Isolated Amplifier including power supply
Spezifikation
Spezifikation
| Produkttyp | Direktbefüllsystem |
| Einwegprodukt | Reusable |