missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AXL, His Tag Recombinant Protein

Artikelnummer. 15647888
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15647888

Marke: Invitrogen™ A31515

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. The purified recombinant protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ assay technologies to identify novel kinase inhibitors. AXL is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen & Z'-LYTE assay technology Available in multiple pack sizes for convenient use Also available in our SelectSceen Kinase Profiling Service. Protein Form: Recombinant, Active, Cytoplasmic, Truncated.

AXL is a chronic myelogenous leukemia-associated oncogene that is also associated with colon cancer and melanoma. The AXL protein is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6, and is involved in the stimulation of cell proliferation. TAM receptors are also involved in phagocytosis of apoptotic cells. Mice lacking all three TAM receptors have several degenerative phenotypes linked to inefficient removal of apoptotic cells and membranes (e.g., in the retina and the male reproductive tract) and develop a severe autoimmune phenotype akin to systemic lupus erythematosus, including the production of broad spectrum auto-antibodies. In addition, Axl may function as a putative entry receptor for filoviruses. Axl is also used to identify a specific subpopulation of human blood dendritic cells, also referred to as Siglec 6+/Axl+ dendritic cells. Cellular expression of Axl can be upregulated by TLR ligands, such as LPS or poly I:C. Soluble Axl is generated by proteolytic cleavage of the membrane form. Increased plasma levels may be indicative of inflammation and cancer.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P30530
Konzentration See Label
Zur Verwendung mit (Anwendung) Kinase Assay
Zusammensetzung 50mM sodium phosphate with 0.1mM PMSF, 300mM NaCl, 25% glycerol, 150mM Imidazole, 0.2mM DTT and no preservative; pH 7
Gen-ID (Entrez) 558
Molekulargewicht 48.3 kDa
Name Human AXL, His Tag
pH-Bereich 7
Reinigungsverfahren Purified
Menge 10 μg
Lagerungsbedingungen -80°C, Avoid Freeze/Thaw Cycles
Kennzeichnung RUO
Gen-Alias Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog
Gebräuchliche Bezeichnung AXL
Gensymbol AXL
Kinase-Familie AXL Kinase Family
Proteinlänge 473-end
Produkttyp Protein
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-tag
Sequenz 473-end
Inhalt und Lagerung -80°C, Avoid Freeze/Thaw Cycles
Versandbedingung Dry Ice
Expressionssystem Insect cells
Proteinfamilie Kinases & Inhibitors
Proteinform Recombinant, Active, Cytoplasmic, Truncated
Form Liquid
Forschungskategorie Signal Transduction
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt