missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B Mouse, Clone: AURKB/1521, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 9212-MSM1-P1
Additional Details : Gewicht : 0.01000kg
Spezifikation
Aurora B | |
Nuclear and Cytoplasmic | |
AURKB/1521 | |
Unconjugated | |
NP_001300884 | |
Mouse | |
39kDa | |
Protein A/G Purified | |
RUO | |
9212 | |
IgG2b |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
AURKB | |
Recombinant human Aurora B protein fragment (aa89-251) | |
HeLa cells. Tonsil, Placenta, Hodgkin's Lymphoma or Colon Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |