missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AURKC (Aurora C) Recombinant Human Protein

AURKC (Aurora C) is a chromosomal passenger protein that forms complexes with AURKB (Aurora B) and inner centromere proteins.

Marke:  Invitrogen™ PR6425A

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15418483

  • 2605.00 CHF / 100 Mikrogramm
Verfügbar ab: 18-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

AURKC (Aurora C) plays a role in organizing microtubules in relation to centrosome/spindle function during mitosis. It is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction.

Spezifikation

Spezifikation

AAH75064
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
AURKC (Aurora C), Aurora A, STK13
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
61.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Aurora Kinase Family
Recombinant
Baculovirus
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote