Learn More
Invitrogen™ AUH Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5109434
Beschreibung
Immunogen sequence: SSEMKTEDEL RVRHLEEENR GIVVLGINRA YGKNSLSKNL IKMLSKAVDA LKSDKKVRTI IIRSEVPGIF CAGADLKERA KMSSSEVGPF VSKIRAVIND IANLPVPTIA AIDGLALGGG LELALACDIR VAASSAKMGL VETKLAIIPG GGGTQRLPRA IGMSLAKELI FSARVLDGKE AKAVGLISHV LEQNQEGDAA YRKALDLARE FLPQGPVAMR VAKLAINQGM EVDLVTGLAI EEACYAQTIP TKDRLEGLLA FKEKRPPRYK GE.
The methylglutaconyl-CoA hydratase, mitochondrial protein binds to the AU-rich element (ARE), a common element found in the 3' UTR of rapidly decaying mRNA such as c-fos, c-myc and granulocyte/ macrophage colony stimulating factor. ARE elements are involved in directing RNA to rapid degradation and deadenylation. AUH is also homologous to enol-CoA hydratase, an enzyme involved in fatty acid degradation, and has been shown to have intrinsic hydratase enzymatic activity. AUH is thus a bifunctional chimera between RNA binding and metabolic enzyme activity. A possible subcellular localization in the mitochondria has been demonstrated for the mouse homolog of this protein which shares 92% identity with the human protein. It has been suggested that AUH may have a novel role as a mitochondrial located AU-binding protein. Human AUH is expressed as a single mRNA species of 1.8 kb, and translated as a 40- kDa precursor protein which is subsequently processed to a 32- kDa mature form.
Spezifikation
| AUH | |
| Polyclonal | |
| Unconjugated | |
| AUH | |
| 3-methylglutaconyl-CoA hydratase; AU RNA binding methylglutaconyl-CoA hydratase; AU RNA binding protein/enoyl-CoA hydratase; AU RNA binding protein/enoyl-coenzyme A hydratase; AU RNA-binding protein/enoyl-Coenzyme A hydratase; AU-binding enoyl-CoA hydratase; AU-binding protein/enoyl-CoA hydratase; Auh; AU-specific RNA-binding enoyl-CoA hydratase; C77140; Itaconyl-CoA hydratase; Methylglutaconyl-CoA hydratase, mitochondrial; muAUH; W91705 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11992, 361215, 549 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13825, Q9JLZ3 | |
| AUH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 68-339 of human AUH (NP_001689.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.