Learn More
Invitrogen™ Attune Xenith Focusing Fluid
Artikelnummer. 31759179
Ansicht ändern
Click to view available options
Menge:
1 Ea.
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
31759179 1 Ea. 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 31759179 Lieferant Invitrogen™ Lieferanten-Nr. X20020
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Attune™ Xenith™ Focusing Fluid is a sterile, pH-buffered isotonic saline solution specifically engineered for use with the Attune Xenith Flow Cytometer. This ready-to-use 1X formulation has been optimized to maintain optimal performance across the complete wavelength range of lasers available on the Attune Xenith system (349 nm - 785 nm).
Attune™ Xenith™ Focusing Fluid is a sterile, pH-buffered isotonic saline solution specifically engineered for use with the Attune Xenith Flow Cytometer. This ready-to-use 1X formulation has been optimized to maintain optimal performance across the complete wavelength range of lasers available on the Attune Xenith system (349 nm - 785 nm), distinguishing it from standard Attune Focusing Fluid, which is designed for the more limited wavelength range of Attune NxT and CytPix systems (405 nm - 637 nm).
The focusing fluid is formulated to minimize bubble formation and prevent cellular material buildup in the flow cell, thereby maintaining clean fluid lines and reducing instrument clogging. Each batch is manufactured under controlled conditions with full lot number traceability and validation for Attune Xenith Flow Cytometer compatibility. The solution is supplied in a convenient 20 L package designed for direct integration with the Attune Xenith fluid cart system, reducing the need for buffer dilution or transfer to alternative containers prior to use.
Features:
  • System Performance Maintenance: Aids optimal operational efficiency and consistent performance parameters
  • Hydrodynamic Focusing Enhancement: Facilitates precise sample positioning and alignment within the flow cell through controlled fluid dynamics, in combination with the acoustic focusing of the system
  • Exceptional Quality Construction: Manufactured to rigorous quality standards to aid reliable performance and reproducibility in laboratory applications
Attune Xenith Focusing Fluid: Optimized Performance Solution for Flow Cytometry Applications
Formulation Assists Consistent System Performance
The Attune Xenith Focusing Fluid represents a specialized solution engineered to maintain optimal system performance through its formulation designed to assist in preserving clean fluid lines within the instrument. This focusing fluid minimizes bubble formation and helps reduce the accumulation of cellular material within the flow cell, thereby reducing the occurrence of instrument clogging that can lead to costly downtime and maintenance requirements.
Ready-to-Use Formulation Streamlines Laboratory Workflow
The focusing fluid is supplied as a ready-to-use 1X concentration, reducing the time-consuming and potentially error-prone dilution steps required by some alternative products. This formulation design reduces preparation time and minimizes the risk of contamination or dilution errors that could affect experimental outcomes. The product is packaged in a convenient 20-liter cube specifically engineered to integrate seamlessly with the Attune Xenith Flow Cytometer's fluid cart system, eliminating the need for buffer transfer to alternative containers and reducing handling requirements.
Rigorous Quality Control Ensures Reliable Performance
Each production batch of Attune Xenith Focusing Fluid undergoes lot testing and the product is verified for use with the Attune Xenith Flow Cytometer platform. The quality assurance program includes complete lot number traceability, enabling researchers to maintain detailed records of reagent provenance for regulatory compliance and experimental documentation. This process helps provide consistent performance characteristics across all batches, providing researchers with the reliability necessary for reproducible experimental results.
Simplified Installation and Operation Protocol
Implementation of the Attune Xenith Focusing Fluid requires minimal setup procedures, with the 20-liter cube designed for direct loading onto the Attune Xenith Fluid cart. The system utilizes the cap and tubing components provided with the instrument, ensuring proper connection and fluid delivery without requiring additional accessories or modifications to the existing flow cytometer configuration.
TRUSTED_SUSTAINABILITY
Spezifikation
Menge 1 Ea.
Zur Verwendung mit (Geräte) Attune Xenith Flow Cytometer
Produkttyp Focusing Fluid Solution
For Research Use Only. Not for use in diagnostic procedures.