Learn More
Thermo Scientific Chemicals Atriale natriuretische Peptide (1-28), Biotin-markiert, Mensch, Schwein
für zell- und molekularbiologische Anwendungen.
Marke: Thermo Scientific Chemicals J66398.M¬
Additional Details : Gewicht : 0.00001kg
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Human, Porcine | |
-30°C to -10°C | |
3306.73 | |
1 mg |
HPLC | |
Lyophilisiertes Pulver | |
Atrial Natriuretic Peptide (1-28), biotin labeled, human, porcine | |
RUO - research use only |
Nur für Forschungszwecke