missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V0D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18274-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c′′, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is known as the D subunit and is found ubiquitously.Spezifikation
ATP6V0D1 | |
Polyclonal | |
Unconjugated | |
ATP6V0D1 | |
32 kDa accessory protein, ATP6D, ATP6DV, ATP6V0D1, P39, V ATPase AC39 subunit, V ATPase subunit d 1, VATX, VMA6, VPATPD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11972, 291969, 9114 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51863, P61421 | |
ATP6V0D1 | |
ATP6V0D1 Fusion Protein Ag13002 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |