Learn More
ATP6V0A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554570
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VQFRDLNPDV NVFQRKFVNE VRRCEEMDRK LRFVEKEIRK ANIPIMDTGE NPEVPFPRDM IDLEANFEKI ENELKEINTN QEALKRNFLE LTELK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
ATP6V0A1 Polyclonal antibody specifically detects ATP6V0A1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
ATP6V0A1 | |
Polyclonal | |
Unconjugated | |
ATP6V0A1 | |
a1; AA959968; adenosine triphosphatase; ATP6a1; ATP6N1; ATP6N1A; Atp6v0a1; ATPase H+ transporting lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa); ATPase H+ transporting V0 subunit a1; ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa); ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1A (110/116kD); ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1A (110/160 kDa); ATPase, H+ transporting, lysosomal non-catalytic accessory protein 1 (110/116kD); ATPase, H+ transporting, lysosomal noncatalytic accessory protein 1a; ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a1; Atpv0a1; Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit; H(+)-transporting two-sector ATPase, 116 kDa accessory protein A1; Stv1; V-A; vacuolar adenosine triphosphatase subunit Ac116; vacuola | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q93050 | |
ATP6V0A1 | |
Recombinant protein corresponding to Human ATP6V0A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only