missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Synthase beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA597483
Additional Details : Gewicht : 0.01000kg
Beschreibung
ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP+ phosphate to ATP. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits. While the F0 consists of a, b, and c subunits.Spezifikation
ATP Synthase beta | |
Polyclonal | |
Unconjugated | |
ATP5F1B | |
ATP synthase F1 subunit beta, ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting mitochondrial F1 complex, alpha subunit, ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, Atp5b, ATP5F1B, ATPMB, ATPSB, beta-subunit, epididymis secretory protein Li 271, f1-ATPase beta, F1-ATPase beta-subunit, F-1-ATPase beta-subunit precursor, fj13e04, fj55c09, HEL-S-271, hm:zehn0534, hypothetical protein LOC554135, im:6793121, MGC5231, mitochondrial ATP synthase beta subunit, mitochondrial ATP synthase subunit beta subunit, mitochondrial ATP synthase, H+ transporting F1 complex beta subunit, mitochondrial ATP synthetase, beta subunit, wu:fj13e04, wu:fj38d01, wu:fj55c09, zgc:111961 | |
Rabbit | |
Protein G | |
RUO | |
11947, 171374, 506 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
P06576, P10719, P56480 | |
ATP5F1B | |
Recombinant Human ATP synthase subunit beta, mitochondrial protein (230-529AA) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |