missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19243-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Dentatorubral pallidoluysian atrophy (DRPLA) is a rare neurodegenerative disorder characterized by cerebellar ataxia, myoclonic epilepsy, choreoathetosis, and dementia. The disorder is related to the expansion from 7-35 copies to 49-93 copies of a trinucleotide repeat (CAG/CAA) within this gene. The encoded protein includes a serine repeat and a region of alternating acidic and basic amino acids, as well as the variable glutamine repeat. Alternative splicing results in two transcripts variants that encode the same protein.Spezifikation
ATN1 | |
Polyclonal | |
Unconjugated | |
ATN1 | |
ATN1, atrophin 1, B37, D12S755E, DRPLA, HRS, NOD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13498, 1822 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35126, P54259 | |
ATN1 | |
ATN1 Fusion Protein Ag5835 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |