Learn More
Astrin Antibody (CL10078) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-88942PE
Artikelnummer. 30295566
  • 884.00 CHF / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Astrin Monoclonal antibody specifically detects Astrin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Astrin
Monoclonal
PE
Astrin, Deepest, hMAP126, MAP126mitotic spindle coiled-coil related protein, mitotic spindle associated protein p126, Mitotic spindle-associated protein p126, sperm associated antigen 5, sperm-associated antigen 5
This antibody was developed using a synthetic peptide derived from Q96R06, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, MAP Kinase Signaling, Stem Cell Markers
10615
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL10078
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen