missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein L4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA562049
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: TSTDQLEALR DILRDITPNV LSFALDFDEA TKMIANDVHT LRRSKATVGR PLIAWRYVPI NVVETLRTRG APTRIVRKVA RNL Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.
The protein encoded by this gene is a member of the apolipoprotein L family and may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver. Two transcript variants encoding two different isoforms have been found for this gene. Only one of the isoforms appears to be a secreted protein.
Spezifikation
| Apolipoprotein L4 | |
| Polyclonal | |
| Unconjugated | |
| APOL4 | |
| APOL4; apolipoprotein L, 4; Apolipoprotein L4; apolipoprotein L-IV; APOLIV; APOL-IV; OTTHUMP00000028937 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BPW4 | |
| APOL4 | |
| Recombinant protein corresponding to Human Apolipoprotein L4. Recombinant protein control fragment (Product #RP-101146). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur