missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA528467
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: Raji, K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
APOL1 is a secreted high density lipoprotein which binds to apolipoprotein A-I. Apolipoprotein A-I is a relatively abundant plasma protein and is the major apoprotein of HDL. It is involved in the formation of most cholesteryl esters in plasma and also promotes efflux of cholesterol from cells. This apolipoprotein L family member may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver.
Spezifikation
| Apolipoprotein L1 | |
| Polyclonal | |
| Unconjugated | |
| APOL1 | |
| APOL; APO-L; APOL- I; APOL1; APOL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L1; Apolipoprotein L-I; FSGS4; OTTHUMP00000028705 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8542 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O14791 | |
| APOL1 | |
| Recombinant protein encompassing a sequence within the center region of human Apolipoprotein L1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur