missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein H Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA582114
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: EPGEEITYSC KPGYVSRGGM RKFICPLTGL WPINTLKCTP RVCPFAGILE NGAVRYTTFE YPNTISFSCN TGFYLNGADS AKCTEEGKWS PELPVCAPII CPPPSIPTFA TLRVYKPSAG NNSLYRDTAV FECLPQHAMF GNDTITC Highest antigen sequence indentity to the following orthologs: Mouse - 74%, Rat - 78%.
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
Spezifikation
| Apolipoprotein H | |
| Polyclonal | |
| Unconjugated | |
| APOH | |
| 2GPI; activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; APOH; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2G1; B2GP1; B2GPI; beta(2)GPI; beta-2-glycoprotein 1; Beta-2-glycoprotein I; beta-2-GPI; beta2-GPI; BG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P02749 | |
| APOH | |
| Recombinant protein corresponding to Human APOH. Recombinant protein control fragment (Product #RP-100506). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur