missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein H Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA547696
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
Spezifikation
| Apolipoprotein H | |
| Polyclonal | |
| Unconjugated | |
| APOH | |
| 2GPI; activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; APOH; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2G1; B2GP1; B2GPI; beta(2)GPI; beta-2-glycoprotein 1; Beta-2-glycoprotein I; beta-2-GPI; beta2-GPI; BG | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 350 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P02749 | |
| APOH | |
| E. coli-derived recombinant human Apolipoprotein H/ApoH Gly20-Cys345. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur