missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E/ApoE Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-41216H
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Apolipoprotein E/ApoE Polyclonal antibody specifically detects Apolipoprotein E/ApoE in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spezifikation
| Apolipoprotein E/ApoE | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human APO-E. The immunogen is located within amino acids 220 - 270 of APO-E. Amino Acid Squence: ARMEEMGSRTRDRLDEVKE | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Cognition and Behavior, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 348 | |
| Store at 4°C in the dark. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur