missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein D Polyclonal Antibody
Goat Polyclonal Antibody
Marke: Invitrogen™ PA537838
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is reported to work in a sandwich-ELISA as both as capture and detection antibody with Product # PA5-37837.
Apolipoprotein D (APOD) is a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism.
Spezifikation
| Apolipoprotein D | |
| Polyclonal | |
| Unconjugated | |
| APOD | |
| AOPDGN; APOD; Apo-D; APO-D protein; Apolipoprotein D | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 347 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P05090 | |
| APOD | |
| Peptide with sequence C-ENGKIKVLNQE, from the internal region of APOD (aa70-80). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur